Product Details

SNP ID
rs200095225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6794817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCTCTCTTGACCTCTTGATTGC[A/G]CAGGCAGTAAATGATGGGATTGAGC
Phenotype
MIM: 608495
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2AG1 PubMed Links

Gene Details

Gene
OR2AG1
Gene Name
olfactory receptor family 2 subfamily AG member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR6A2
Gene Name
olfactory receptor family 6 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003696.2 1092 Missense Mutation CGC,TGC R298C NP_003687.2

View Full Product Details