Product Details

SNP ID
rs200120334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:69295710 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCTCTGTCTCTCCCAGGCCCTGC[A/G]TGTTGCGGTGAGAGGAGCATTTGTG
Phenotype
MIM: 605625
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYEOV PubMed Links

Gene Details

Gene
MYEOV
Gene Name
myeloma overexpressed
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293291.1 558 Missense Mutation CAT,CGT H87R NP_001280220.1
NM_001293294.1 558 Missense Mutation CAT,CGT H29R NP_001280223.1
NM_001293296.1 558 Missense Mutation CAT,CGT H29R NP_001280225.1
NM_001300923.1 558 Missense Mutation CAT,CGT H29R NP_001287852.1
NM_001300924.1 558 Missense Mutation CAT,CGT H29R NP_001287853.1
NM_138768.3 558 Missense Mutation CAT,CGT H87R NP_620123.2

View Full Product Details