Product Details

SNP ID
rs199984603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:372709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTTTCCTGGGGGGGCTGGGAG[C/G]CTGCCACTGAACTTCACCCATCAGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
B4GALNT4 PubMed Links

Gene Details

Gene
B4GALNT4
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178537.4 312 Missense Mutation AGC,AGG S101R NP_848632.2
XM_017017654.1 312 Missense Mutation AGC,AGG S9R XP_016873143.1

View Full Product Details