Product Details

SNP ID
rs199988558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35663699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATTGCCAGGAAGATAGGCAGCT[C/T]ATCTGTGTCCTGTGTCCAGTCATTG
Phenotype
MIM: 612298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM44 PubMed Links

Gene Details

Gene
TRIM44
Gene Name
tripartite motif containing 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017583.5 948 Silent Mutation CTC,CTT L196L NP_060053.2
XM_006718254.1 948 Silent Mutation CTC,CTT L196L XP_006718317.1

View Full Product Details