Product Details

SNP ID
rs200355216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:111543937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTCTCTAGGGCAGCCAGTCTGC[C/T]GGGGAGGGACACAGAGGCCTTGTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf88 PubMed Links

Gene Details

Gene
C11orf88
Gene Name
chromosome 11 open reading frame 88
There are no transcripts associated with this gene.

Gene
LAYN
Gene Name
layilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258390.1 460 Missense Mutation CGG,TGG R42W NP_001245319.1
NM_001258391.1 460 UTR 5 NP_001245320.1
NM_001318799.1 460 UTR 5 NP_001305728.1
NM_178834.4 460 Missense Mutation CGG,TGG R34W NP_849156.1
XM_006718769.3 460 UTR 5 XP_006718832.1
XM_006718770.3 460 UTR 5 XP_006718833.1

View Full Product Details