Product Details

SNP ID
rs200465752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:101922708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGATATTACTGCAGCAACAAAAA[A/T]TATGTCGAAATCGAGCACGTAAATA
Phenotype
MIM: 607666 MIM: 614634
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANGPTL5 PubMed Links

Gene Details

Gene
ANGPTL5
Gene Name
angiopoietin like 5
There are no transcripts associated with this gene.

Gene
CEP126
Gene Name
centrosomal protein 126
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020802.3 197 Missense Mutation ATA,TTA I66L NP_065853.3
XM_005271627.4 197 Missense Mutation ATA,TTA I66L XP_005271684.2

View Full Product Details