Product Details

SNP ID
rs199762736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124870273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCCCCACCTCGTCTACGCTGGC[A/G]CAAGGAGGATGGGGAACTGCCCACA
Phenotype
MIM: 608630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ROBO3 PubMed Links

Gene Details

Gene
ROBO3
Gene Name
roundabout guidance receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022370.3 1044 Missense Mutation CAC,CGC H292R NP_071765.2
XM_017018122.1 1044 Missense Mutation CAC,CGC H270R XP_016873611.1

View Full Product Details