Product Details

SNP ID
rs199607712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56375785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTACTCATTAGAGCGGATACAAG[A/T]CTCAACACACCAATGTACTTCTTTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR8U1 PubMed Links

Gene Details

Gene
OR8U1
Gene Name
olfactory receptor family 8 subfamily U member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005204.1 162 Missense Mutation AGA,AGT R54S NP_001005204.1

View Full Product Details