Product Details
- SNP ID
-
rs199724415
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:125747714 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTGGCCAAGTGATAGAAATTGTT[C/T]AGTGTAGGATGTGCCACCTCCAGTT
- Phenotype
-
MIM: 606861
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PATE1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2114084,rs537916] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PATE1
- Gene Name
- prostate and testis expressed 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138294.2 |
151 |
Nonsense Mutation |
CAG,TAG |
Q47* |
NP_612151.1 |
View Full Product Details