Product Details

SNP ID
rs201548958
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55554428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTGTCCTAATTGCAGACTAT[A/G]CATGATCCCTGTTGGAGCTTTCATC
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR4C15 PubMed Links

Gene Details

Gene
OR4C15
Gene Name
olfactory receptor family 4 subfamily C member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001920.1 122 Missense Mutation TAC,TGC Y41C NP_001001920.1

View Full Product Details