Product Details

SNP ID
rs201582713
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47240175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCAGGAGGCTGGGCCTGCATCC[A/G]GAAGAGGACGGTGAGGAGCAGCACT
Phenotype
MIM: 171650 MIM: 600811 MIM: 602423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACP2 PubMed Links

Gene Details

Gene
ACP2
Gene Name
acid phosphatase 2, lysosomal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302489.1 1200 Missense Mutation NP_001289418.1
NM_001302490.1 1200 Missense Mutation NP_001289419.1
NM_001302491.1 1200 Missense Mutation NP_001289420.1
NM_001302492.1 1200 Missense Mutation NP_001289421.1
NM_001610.3 1200 Missense Mutation NP_001601.1
Gene
DDB2
Gene Name
damage specific DNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000107.2 1200 Intron NP_000098.1
NM_001300734.1 1200 Intron NP_001287663.1
Gene
NR1H3
Gene Name
nuclear receptor subfamily 1 group H member 3
There are no transcripts associated with this gene.

View Full Product Details