Product Details

SNP ID
rs201516616
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:73958409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCAAATAGCAGAGGCCTACGACG[C/T]GCTGAGTGACCGTGAGTAGGTGTGG
Phenotype
MIM: 610263
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB13 PubMed Links

Gene Details

Gene
DNAJB13
Gene Name
DnaJ heat shock protein family (Hsp40) member B13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153614.3 1169 Missense Mutation GCG,GTG A54V NP_705842.2
XM_005273984.3 1169 Missense Mutation GCG,GTG A54V XP_005274041.1
XM_011545004.2 1169 Missense Mutation GCG,GTG A88V XP_011543306.1
XM_011545005.2 1169 Missense Mutation GCG,GTG A88V XP_011543307.1
XM_011545007.2 1169 UTR 5 XP_011543309.1
XM_011545009.2 1169 UTR 5 XP_011543311.1
XM_011545013.2 1169 Missense Mutation GCG,GTG A88V XP_011543315.1
XM_011545014.2 1169 Missense Mutation GCG,GTG A88V XP_011543316.1
XM_011545015.2 1169 Intron XP_011543317.1
XM_017017675.1 1169 UTR 5 XP_016873164.1
XM_017017676.1 1169 UTR 5 XP_016873165.1
XM_017017677.1 1169 UTR 5 XP_016873166.1
XM_017017678.1 1169 UTR 5 XP_016873167.1
XM_017017679.1 1169 Missense Mutation GCG,GTG A88V XP_016873168.1

View Full Product Details