Product Details

SNP ID
rs201471226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58118934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAGAAGAAGTTTATTTGATTGT[C/T]CTTACAGAAGGAGAGGGTGAAGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR9I1 PubMed Links

Gene Details

Gene
OR9I1
Gene Name
olfactory receptor family 9 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005211.1 511 Missense Mutation AAC,GAC N171D NP_001005211.1
Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005212.3 511 Intron NP_001005212.1

View Full Product Details