Product Details

SNP ID
rs201636126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:77102954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCTGAAGCAGCGCGGGGAGAAG[C/T]GCCAGGATGGGGAGAAGCTGCTGCA
Phenotype
MIM: 602537 MIM: 164340
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAPN5 PubMed Links

Gene Details

Gene
CAPN5
Gene Name
calpain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004055.4 115 Intron NP_004046.2
XM_011545225.1 115 Intron XP_011543527.1
XM_017018223.1 115 Intron XP_016873712.1
Gene
OMP
Gene Name
olfactory marker protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006189.1 115 Missense Mutation CGC,TGC R39C NP_006180.1

View Full Product Details