Product Details

SNP ID
rs202160057
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5269586 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGCACCTTCTTGCCATGGGCCTT[G/T]ACCTTGGGGTTGCCCAGGATGGCAG
Phenotype
MIM: 142100
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
HBE1 PubMed Links

Gene Details

Gene
HBE1
Gene Name
hemoglobin subunit epsilon 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005330.3 436 Silent Mutation GTA,GTC V61V NP_005321.1

View Full Product Details