Product Details

SNP ID
rs201085695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4803573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGTGTACAACTCGGGGCACAT[C/T]ATGGCCAAAGCGGTAGGTGAGGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52R1 PubMed Links

Gene Details

Gene
OR52R1
Gene Name
olfactory receptor family 52 subfamily R member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005177.3 808 Missense Mutation AAT,GAT N270D NP_001005177.3

View Full Product Details