Product Details

SNP ID
rs201223736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4489571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATCCTGTCTTATGTCTTCATCC[C/T]TCAGGCAGTTCTCCAGCTTGCCTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52K1 PubMed Links

Gene Details

Gene
OR52K1
Gene Name
olfactory receptor family 52 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005171.2 693 Missense Mutation CCT,CTT P224L NP_001005171.2

View Full Product Details