Product Details

SNP ID
rs201135328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66437121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACTTACTCCTTCACCACGCGAAT[A/G]CCCAGAGAACGGGCAGACCCGATGA
Phenotype
MIM: 611826
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL11 PubMed Links

Gene Details

Gene
MRPL11
Gene Name
mitochondrial ribosomal protein L11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016050.4 550 Silent Mutation NP_057134.1
NM_170738.3 550 Silent Mutation NP_733934.1
NM_170739.3 550 Silent Mutation NP_733935.1
XM_017018135.1 550 Silent Mutation XP_016873624.1

View Full Product Details