Product Details

SNP ID
rs201410689
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65896828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTACCTGTTCACAAGGCCTTCGA[C/T]GTACCCCTGGAGGCGGCCCCAGGGC
Phenotype
MIM: 605360 MIM: 608296 MIM: 136515
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC85B PubMed Links

Gene Details

Gene
CCDC85B
Gene Name
coiled-coil domain containing 85B
There are no transcripts associated with this gene.

Gene
FIBP
Gene Name
FGF1 intracellular binding protein
There are no transcripts associated with this gene.

Gene
FOSL1
Gene Name
FOS like 1, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300844.1 465 Missense Mutation NP_001287773.1
NM_001300855.1 465 Missense Mutation NP_001287784.1
NM_001300856.1 465 Intron NP_001287785.1
NM_001300857.1 465 Intron NP_001287786.1
NM_005438.4 465 Missense Mutation NP_005429.1

View Full Product Details