Product Details

SNP ID
rs201204707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64827114 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCACGTGTACTAGGTGGTTGAAG[C/T]TGGTAGGCGGCGAGATGAGCTTGGA
Phenotype
MIM: 613991
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC42BPG PubMed Links

Gene Details

Gene
CDC42BPG
Gene Name
CDC42 binding protein kinase gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017525.2 4474 Missense Mutation AAC,AGC N1442S NP_059995.2
XM_011545155.2 4474 Missense Mutation AAC,AGC N1460S XP_011543457.1
XM_011545156.2 4474 Missense Mutation AAC,AGC N1453S XP_011543458.1
XM_011545157.2 4474 Silent Mutation CAA,CAG Q1427Q XP_011543459.1
XM_011545158.2 4474 Silent Mutation CAA,CAG Q1427Q XP_011543460.1
XM_011545159.2 4474 Missense Mutation AAC,AGC N1460S XP_011543461.1
XM_011545160.2 4474 Missense Mutation AAC,AGC N1362S XP_011543462.1
XM_011545161.2 4474 Intron XP_011543463.1
XM_017017996.1 4474 Missense Mutation AAC,AGC N1459S XP_016873485.1
XM_017017997.1 4474 Intron XP_016873486.1
XM_017017998.1 4474 Intron XP_016873487.1
XM_017017999.1 4474 Missense Mutation AAC,AGC N756S XP_016873488.1

View Full Product Details