Product Details

SNP ID
rs201162064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64925862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTCCGCAGAGCCCGGCCCCGCC[A/G]CTCCCACAGCTCCTCTCAGTTCCGC
Phenotype
MIM: 616225 MIM: 609651 MIM: 601644
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATG2A PubMed Links

Gene Details

Gene
ATG2A
Gene Name
autophagy related 2A
There are no transcripts associated with this gene.

Gene
GPHA2
Gene Name
glycoprotein hormone alpha 2
There are no transcripts associated with this gene.

Gene
PPP2R5B
Gene Name
protein phosphatase 2 regulatory subunit B'beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006244.3 183 Missense Mutation CAC,CGC H43R NP_006235.1
XM_011545132.2 183 Missense Mutation CAC,CGC H14R XP_011543434.1
XM_011545133.1 183 UTR 5 XP_011543435.1
XM_011545134.2 183 UTR 5 XP_011543436.1

View Full Product Details