Product Details

SNP ID
rs201118576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4769124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCTGGGGGCTGACCGACCATAG[C/T]GATACACCAAGGACAGGCTCAGCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR51F1 PubMed Links

Gene Details

Gene
OR51F1
Gene Name
olfactory receptor family 51 subfamily F member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004752.1 794 Missense Mutation CAC,CGC H265R NP_001004752.1

View Full Product Details