Product Details
- SNP ID
-
rs201400802
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:68158361 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGCACAGTCTGTGTAGCTCACA[A/G]GCACGCCCACAGTGCCACTGTGCTC
- Phenotype
-
MIM: 610881
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KMT5B
PubMed Links
Gene Details
- Gene
- KMT5B
- Gene Name
- lysine methyltransferase 5B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001300907.1 |
2337 |
Missense Mutation |
CCT,CTT |
P490L |
NP_001287836.1 |
NM_001300908.1 |
2337 |
Missense Mutation |
CCT,CTT |
P422L |
NP_001287837.1 |
NM_001300909.1 |
2337 |
Intron |
|
|
NP_001287838.1 |
NM_016028.4 |
2337 |
Intron |
|
|
NP_057112.3 |
NM_017635.4 |
2337 |
Missense Mutation |
CCT,CTT |
P662L |
NP_060105.3 |
XM_005274035.3 |
2337 |
Missense Mutation |
CCT,CTT |
P662L |
XP_005274092.2 |
XM_005274036.3 |
2337 |
Missense Mutation |
CCT,CTT |
P639L |
XP_005274093.2 |
XM_006718581.1 |
2337 |
Missense Mutation |
CCT,CTT |
P639L |
XP_006718644.1 |
XM_011545091.1 |
2337 |
Missense Mutation |
CCT,CTT |
P662L |
XP_011543393.1 |
XM_011545092.2 |
2337 |
Missense Mutation |
CCT,CTT |
P591L |
XP_011543394.1 |
XM_011545093.2 |
2337 |
Missense Mutation |
CCT,CTT |
P248L |
XP_011543395.1 |
XM_011545094.2 |
2337 |
Intron |
|
|
XP_011543396.1 |
XM_017017876.1 |
2337 |
Missense Mutation |
CCT,CTT |
P490L |
XP_016873365.1 |
XM_017017877.1 |
2337 |
Missense Mutation |
CCT,CTT |
P490L |
XP_016873366.1 |
XM_017017878.1 |
2337 |
Missense Mutation |
CCT,CTT |
P490L |
XP_016873367.1 |
XM_017017879.1 |
2337 |
Missense Mutation |
CCT,CTT |
P490L |
XP_016873368.1 |
XM_017017880.1 |
2337 |
Intron |
|
|
XP_016873369.1 |
View Full Product Details