Product Details

SNP ID
rs201091442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2797774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATTGCCATAGCCACCATGAAGG[A/T]GGGGGAGGTGTGCCACATCACCTGC
Phenotype
MIM: 600611
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FKBP4 PubMed Links

Gene Details

Gene
FKBP4
Gene Name
FK506 binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002014.3 494 Missense Mutation GAG,GTG E99V NP_002005.1
XM_011520929.2 494 Missense Mutation GAG,GTG E54V XP_011519231.1

View Full Product Details