Product Details

SNP ID
rs201115819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9068811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTACTGGGACATCTTGCAGAACC[A/G]TGAAGAACAAGCTCAGTGTCTGATT
Phenotype
MIM: 103950 MIM: 604874
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
A2M PubMed Links

Gene Details

Gene
A2M
Gene Name
alpha-2-macroglobulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000014.4 4408 Missense Mutation ACG,ATG T1432M NP_000005.2
XM_006719056.2 4408 Missense Mutation ACG,ATG T1432M XP_006719119.1
Gene
A2M-AS1
Gene Name
A2M antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
KLRG1
Gene Name
killer cell lectin like receptor G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005810.3 4408 Intron NP_005801.3
XM_017018682.1 4408 Intron XP_016874171.1
XM_017018683.1 4408 Intron XP_016874172.1
XM_017018684.1 4408 Intron XP_016874173.1
XM_017018685.1 4408 Intron XP_016874174.1
Gene
LINC00612
Gene Name
long intergenic non-protein coding RNA 612
There are no transcripts associated with this gene.

View Full Product Details