Product Details
- SNP ID
-
rs200979804
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:71611816 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGTGGCCAGGCATGTTGGGATA[C/T]TATATGTAAACATCTTGGCTTGAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZFC3H1
PubMed Links
Gene Details
- Gene
- ZFC3H1
- Gene Name
- zinc finger C3H1-type containing
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144982.4 |
6058 |
Missense Mutation |
AAT,AGT |
N1900S |
NP_659419.3 |
View Full Product Details