Product Details
- SNP ID
-
rs201054359
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52316020 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTCTTGCTTGGCCTTCTGCAGGG[C/G]GCCCTCCAGCTCGGCCAGCTTGCAG
- Phenotype
-
MIM: 602765
MIM: 601928
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT83
PubMed Links
Gene Details
- Gene
- KRT83
- Gene Name
- keratin 83
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002282.3 |
1198 |
Missense Mutation |
CCC,GCC |
P379A |
NP_002273.3 |
- Gene
- KRT86
- Gene Name
- keratin 86
There are no transcripts associated with this gene.
View Full Product Details