Product Details

SNP ID
rs200819983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53192337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCCCTGCCCACTTACTTTCTTG[C/G]GGTCTCACTCTGAGCACGACCGTGC
Phenotype
MIM: 147559
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITGB7 PubMed Links

Gene Details

Gene
ITGB7
Gene Name
integrin subunit beta 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000889.2 2380 Silent Mutation CCC,CCG P716P NP_000880.1
XM_005268851.3 2380 Silent Mutation CCC,CCG P716P XP_005268908.1
XM_005268852.4 2380 Silent Mutation CCC,CCG P716P XP_005268909.1
XM_006719376.3 2380 Silent Mutation CCC,CCG P716P XP_006719439.1
Gene
ZNF740
Gene Name
zinc finger protein 740
There are no transcripts associated with this gene.

View Full Product Details