Product Details

SNP ID
rs200095319
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2797752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGAGGTCATCAAGGCTTGGGAC[A/C]TTGCCATAGCCACCATGAAGGTGGG
Phenotype
MIM: 600611
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FKBP4 PubMed Links

Gene Details

Gene
FKBP4
Gene Name
FK506 binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002014.3 472 Missense Mutation ATT,CTT I92L NP_002005.1
XM_011520929.2 472 Missense Mutation ATT,CTT I47L XP_011519231.1

View Full Product Details