Product Details

SNP ID
rs200050129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27028418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATATTGCACAGTCACAGCTGA[A/G]GACAAGAAGTGGTACCCATAGCCAG
Phenotype
MIM: 603800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MED21 PubMed Links

Gene Details

Gene
MED21
Gene Name
mediator complex subunit 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271811.1 448 UTR 3 NP_001258740.1
NM_004264.4 448 Missense Mutation AAG,AGG K131R NP_004255.2

View Full Product Details