Product Details

SNP ID
rs200445671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113299946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGCCCCAGGGCGCCTGCCAGG[A/G]CCCACACCAGCAGTCCGTCTGGCTC
Phenotype
MIM: 609841 MIM: 609666
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6762 PubMed Links

Gene Details

Gene
MIR6762
Gene Name
microRNA 6762
There are no transcripts associated with this gene.

Gene
SLC8B1
Gene Name
solute carrier family 8 member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006719607.2 2200 Missense Mutation GCC,GTC A334V XP_006719670.1
XM_011538749.2 2200 Missense Mutation GCC,GTC A529V XP_011537051.1
XM_011538750.2 2200 Missense Mutation GCC,GTC A473V XP_011537052.1
XM_011538752.2 2200 Missense Mutation GCC,GTC A334V XP_011537054.1
XM_017019978.1 2200 Intron XP_016875467.1
Gene
TPCN1
Gene Name
two pore segment channel 1
There are no transcripts associated with this gene.

View Full Product Details