Product Details
- SNP ID
-
rs200446552
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:122505608 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGTGTGGGTTTGTGATCTTGCCAT[C/T]TCATACTGAGCATCTAAATTTCGAA
- Phenotype
-
MIM: 616381
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RSRC2
PubMed Links
Gene Details
- Gene
- RSRC2
- Gene Name
- arginine and serine rich coiled-coil 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_023012.5 |
1297 |
Silent Mutation |
GAA,GAG |
E408E |
NP_075388.2 |
XM_005253601.1 |
1297 |
Silent Mutation |
GAA,GAG |
E409E |
XP_005253658.1 |
XM_005253602.2 |
1297 |
Silent Mutation |
GAA,GAG |
E361E |
XP_005253659.1 |
XM_005253604.1 |
1297 |
Silent Mutation |
GAA,GAG |
E350E |
XP_005253661.1 |
XM_011538688.1 |
1297 |
Silent Mutation |
GAA,GAG |
E379E |
XP_011536990.1 |
XM_011538689.1 |
1297 |
Silent Mutation |
GAA,GAG |
E350E |
XP_011536991.1 |
XM_017019830.1 |
1297 |
Silent Mutation |
GAA,GAG |
E378E |
XP_016875319.1 |
XM_017019831.1 |
1297 |
Silent Mutation |
GAA,GAG |
E361E |
XP_016875320.1 |
XM_017019832.1 |
1297 |
Silent Mutation |
GAA,GAG |
E177E |
XP_016875321.1 |
XM_017019833.1 |
1297 |
Silent Mutation |
GAA,GAG |
E176E |
XP_016875322.1 |
- Gene
- ZCCHC8
- Gene Name
- zinc finger CCHC-type containing 8
There are no transcripts associated with this gene.
View Full Product Details