Product Details
- SNP ID
-
rs200771163
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:22449793 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAACTTCGCCTTCAGTACATGATGA[C/T]TGGCAGTTGGTAGTAGGTTGCTGAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C2CD5
PubMed Links
Gene Details
- Gene
- C2CD5
- Gene Name
- C2 calcium dependent domain containing 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286173.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1043Q |
NP_001273102.1 |
NM_001286174.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1041Q |
NP_001273103.1 |
NM_001286175.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1044Q |
NP_001273104.1 |
NM_001286176.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1041Q |
NP_001273105.1 |
NM_001286177.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1032Q |
NP_001273106.1 |
NM_014802.2 |
2394 |
Silent Mutation |
CAA,CAG |
Q990Q |
NP_055617.1 |
XM_005253538.2 |
2394 |
Silent Mutation |
CAA,CAG |
Q981Q |
XP_005253595.1 |
XM_005253539.3 |
2394 |
Silent Mutation |
CAA,CAG |
Q663Q |
XP_005253596.1 |
XM_006719170.2 |
2394 |
Silent Mutation |
CAA,CAG |
Q1007Q |
XP_006719233.1 |
XM_017020264.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1151Q |
XP_016875753.1 |
XM_017020265.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1141Q |
XP_016875754.1 |
XM_017020266.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1134Q |
XP_016875755.1 |
XM_017020267.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1127Q |
XP_016875756.1 |
XM_017020268.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1105Q |
XP_016875757.1 |
XM_017020269.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1100Q |
XP_016875758.1 |
XM_017020270.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1087Q |
XP_016875759.1 |
XM_017020271.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1078Q |
XP_016875760.1 |
XM_017020272.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1054Q |
XP_016875761.1 |
XM_017020273.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1037Q |
XP_016875762.1 |
XM_017020274.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1031Q |
XP_016875763.1 |
XM_017020275.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1027Q |
XP_016875764.1 |
XM_017020276.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1022Q |
XP_016875765.1 |
XM_017020277.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1015Q |
XP_016875766.1 |
XM_017020278.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q1003Q |
XP_016875767.1 |
XM_017020279.1 |
2394 |
Silent Mutation |
CAA,CAG |
Q769Q |
XP_016875768.1 |
View Full Product Details