Product Details

SNP ID
rs199705102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53253518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAAACTGGGGGGAGAACTATGACC[A/G]GCAGCGTGCCTTCTCAAGGACCTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MFSD5 PubMed Links

Gene Details

Gene
MFSD5
Gene Name
major facilitator superfamily domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170790.1 828 Missense Mutation CAG,CGG Q335R NP_001164261.1
NM_032889.4 828 Missense Mutation CAG,CGG Q228R NP_116278.3
XM_005269197.1 828 Missense Mutation CAG,CGG Q335R XP_005269254.1
XM_005269198.4 828 Missense Mutation CAG,CGG Q228R XP_005269255.1

View Full Product Details