Product Details

SNP ID
rs199634350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6867556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGACACTGACCTTCAGCGCCTCG[A/G]CTCCAGCGCCATGGCGCCCTCCAGG
Phenotype
MIM: 611658 MIM: 190450 MIM: 601447
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105369632 PubMed Links

Gene Details

Gene
LOC105369632
Gene Name
uncharacterized LOC105369632
There are no transcripts associated with this gene.

Gene
SPSB2
Gene Name
splA/ryanodine receptor domain and SOCS box containing 2
There are no transcripts associated with this gene.

Gene
TPI1
Gene Name
triosephosphate isomerase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000365.5 137 UTR 5 NP_000356.1
NM_001159287.1 137 Missense Mutation GAC,GGC D34G NP_001152759.1
NM_001258026.1 137 Intron NP_001244955.1
Gene
USP5
Gene Name
ubiquitin specific peptidase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098536.1 137 Intron NP_001092006.1
NM_003481.2 137 Intron NP_003472.2

View Full Product Details