Product Details

SNP ID
rs199803671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109279692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCAAAGCAGGGCTATAGGCTT[G/T]TTCCCCTGTGCACCCAGGTACTGGG
Phenotype
MIM: 609429
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FOXN4 PubMed Links

Gene Details

Gene
FOXN4
Gene Name
forkhead box N4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213596.2 1827 Missense Mutation AAA,AAC K511N NP_998761.2
XM_011537922.2 1827 Missense Mutation AAA,AAC K512N XP_011536224.1
XM_011537923.2 1827 Intron XP_011536225.1
XM_017018818.1 1827 Missense Mutation AAA,AAC K468N XP_016874307.1
XM_017018819.1 1827 Intron XP_016874308.1

View Full Product Details