Product Details

SNP ID
rs199827127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55837229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTTGAGGCGCCAGTAGACTTTG[C/T]CCTTGAAGAAGTAGACTCGGCCATC
Phenotype
MIM: 606092 MIM: 601807
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC14 PubMed Links

Gene Details

Gene
DNAJC14
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
There are no transcripts associated with this gene.

Gene
MMP19
Gene Name
matrix metallopeptidase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272101.1 1227 UTR 3 NP_001259030.1
NM_002429.5 1227 Missense Mutation GAC,GGC D445G NP_002420.1
XM_006719401.3 1227 Missense Mutation GAC,GGC D216G XP_006719464.1
XM_011538359.1 1227 Missense Mutation GAC,GGC D363G XP_011536661.1
XM_017019308.1 1227 Intron XP_016874797.1
XM_017019309.1 1227 UTR 3 XP_016874798.1
Gene
TMEM198B
Gene Name
transmembrane protein 198B (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details