Product Details

SNP ID
rs199900632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:32792668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTTGTAGGCATGATGCAGTTC[C/T]GTGTGTGCCCACAGAGAATACAGAA
Phenotype
MIM: 602861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKP2 PubMed Links

Gene Details

Gene
PKP2
Gene Name
plakophilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005242.2 2668 Silent Mutation ACA,ACG T807T NP_001005242.2
NM_004572.3 2668 Silent Mutation ACA,ACG T851T NP_004563.2

View Full Product Details