Product Details

SNP ID
rs199903998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109388834 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTCCGCAAGCGTTTCAGCGAGA[A/G]CCTCATATACGTAACGTGACCCACT
Phenotype
MIM: 614636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO1H PubMed Links

Gene Details

Gene
MYO1H
Gene Name
myosin IH
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101421.3 558 Missense Mutation AAC,AGC N39S NP_001094891.3
XM_011538223.2 558 Missense Mutation AAC,AGC N39S XP_011536525.1
XM_017019207.1 558 Missense Mutation AAC,AGC N39S XP_016874696.1

View Full Product Details