Product Details

SNP ID
rs199738753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52424672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACTGGTGGTGAAGGAGTAGC[C/T]GCTGCCCCCACCGAGGCCCAGGTTT
Phenotype
MIM: 609025
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KRT75 PubMed Links

Gene Details

Gene
KRT75
Gene Name
keratin 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004693.2 1523 Missense Mutation AGC,GGC S501G NP_004684.2

View Full Product Details