Product Details

SNP ID
rs199673753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109855619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGGGCTCATGGGGGTGGCTCAC[C/T]TGGAAGATCTTCTGCACGATCCAGC
Phenotype
MIM: 608949
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLTP PubMed Links

Gene Details

Gene
GLTP
Gene Name
glycolipid transfer protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016433.3 Intron NP_057517.1
XM_011538434.2 Intron XP_011536736.1

View Full Product Details