Product Details
- SNP ID
-
rs199654159
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:110619912 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCTGTGATCCCGACTGCAGCTC[C/T]GTGGATTTCAGTGTCTTTTCTGCCT
- Phenotype
-
MIM: 609863
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TCTN1
PubMed Links
Gene Details
- Gene
- TCTN1
- Gene Name
- tectonic family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001082537.2 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
NP_001076006.1 |
NM_001082538.2 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
NP_001076007.1 |
NM_001173975.2 |
453 |
Silent Mutation |
TCC,TCT |
S43S |
NP_001167446.1 |
NM_001173976.1 |
453 |
Silent Mutation |
TCC,TCT |
S39S |
NP_001167447.1 |
NM_001319680.1 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
NP_001306609.1 |
NM_001319681.1 |
453 |
UTR 5 |
|
|
NP_001306610.1 |
NM_001319682.1 |
453 |
Silent Mutation |
TCC,TCT |
S43S |
NP_001306611.1 |
NM_024549.5 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
NP_078825.2 |
XM_005253934.3 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
XP_005253991.1 |
XM_005253935.3 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
XP_005253992.1 |
XM_006719594.2 |
453 |
Silent Mutation |
TCC,TCT |
S43S |
XP_006719657.1 |
XM_006719595.2 |
453 |
UTR 5 |
|
|
XP_006719658.1 |
XM_006719596.2 |
453 |
UTR 5 |
|
|
XP_006719659.1 |
XM_006719597.3 |
453 |
Intron |
|
|
XP_006719660.1 |
XM_006719598.2 |
453 |
UTR 5 |
|
|
XP_006719661.1 |
XM_006719599.2 |
453 |
UTR 5 |
|
|
XP_006719662.1 |
XM_006719600.2 |
453 |
Intron |
|
|
XP_006719663.1 |
XM_011538733.2 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
XP_011537035.1 |
XM_011538734.2 |
453 |
Intron |
|
|
XP_011537036.1 |
XM_011538735.1 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
XP_011537037.1 |
XM_011538737.2 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
XP_011537039.1 |
XM_011538738.2 |
453 |
Silent Mutation |
TCC,TCT |
S99S |
XP_011537040.1 |
XM_017019964.1 |
453 |
Silent Mutation |
TCC,TCT |
S43S |
XP_016875453.1 |
XM_017019965.1 |
453 |
UTR 5 |
|
|
XP_016875454.1 |
XM_017019966.1 |
453 |
UTR 5 |
|
|
XP_016875455.1 |
XM_017019967.1 |
453 |
UTR 5 |
|
|
XP_016875456.1 |
XM_017019968.1 |
453 |
Intron |
|
|
XP_016875457.1 |
XM_017019969.1 |
453 |
UTR 5 |
|
|
XP_016875458.1 |
View Full Product Details