Product Details
- SNP ID
-
rs201678034
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:32107493 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACCCTCAAACAGCAGTATGATGA[A/G]CTGGAGGCTGAGTACGACAGCCTCA
- Phenotype
-
MIM: 602204
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BICD1
PubMed Links
Gene Details
- Gene
- BICD1
- Gene Name
- BICD cargo adaptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001003398.1 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
NP_001003398.1 |
NM_001714.2 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
NP_001705.2 |
XM_005253465.3 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_005253522.1 |
XM_005253466.2 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_005253523.1 |
XM_006719134.3 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_006719197.1 |
XM_011520812.2 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_011519114.1 |
XM_011520813.1 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_011519115.1 |
XM_011520814.2 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_011519116.1 |
XM_011520815.2 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_011519117.1 |
XM_011520816.2 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_011519118.1 |
XM_017019797.1 |
243 |
Silent Mutation |
GAA,GAG |
E54E |
XP_016875286.1 |
View Full Product Details