Product Details

SNP ID
rs201819683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120099154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCCCGTGGGTCCCCCGATAATA[C/T]CTGCAGGGCCAGAGTGTGCGGCAGC
Phenotype
MIM: 617002 MIM: 604199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BICDL1 PubMed Links

Gene Details

Gene
BICDL1
Gene Name
BICD family like cargo adaptor 1
There are no transcripts associated with this gene.

Gene
RAB35
Gene Name
RAB35, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167606.1 319 Silent Mutation ACA,ACG T76T NP_001161078.1
NM_006861.6 319 Silent Mutation ACA,ACG T76T NP_006852.1
XM_011537779.1 319 UTR 5 XP_011536081.1
XM_017018723.1 319 UTR 5 XP_016874212.1

View Full Product Details