Product Details

SNP ID
rs201810021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:107734339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTTCTTCCTCCTCTGGTGCTGA[C/T]GAACTGGAGGTGGGCCCTTTGCAGG
Phenotype
MIM: 605780
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929162 PubMed Links

Gene Details

Gene
LOC101929162
Gene Name
uncharacterized LOC101929162
There are no transcripts associated with this gene.

Gene
PRDM4
Gene Name
PR domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012406.3 2714 Silent Mutation TCC,TCT S759S NP_036538.3
XM_005268593.1 2714 Silent Mutation XP_005268650.1

View Full Product Details