Product Details

SNP ID
rs202089444
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57235789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTCAGGCGGTTCCAGGGCTCC[A/G]GGTTGTTCTTTCTGTCCCAGCTGTG
Phenotype
MIM: 604637 MIM: 138450 MIM: 615521
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NDUFA4L2 PubMed Links
Additional Information
For this assay, SNP(s) [rs58212469] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
NDUFA4L2
Gene Name
NDUFA4, mitochondrial complex associated like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020142.3 1046 Missense Mutation CCG,CTG P56L NP_064527.1
XM_005269033.2 1046 Missense Mutation CCG,CTG P130L XP_005269090.1
XM_011538573.2 1046 Missense Mutation CCG,CTG P134L XP_011536875.1
Gene
NXPH4
Gene Name
neurexophilin 4
There are no transcripts associated with this gene.

Gene
SHMT2
Gene Name
serine hydroxymethyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166356.1 1046 Intron NP_001159828.1
NM_001166357.1 1046 Intron NP_001159829.1
NM_001166358.1 1046 Intron NP_001159830.1
NM_001166359.1 1046 Intron NP_001159831.1
NM_005412.5 1046 Intron NP_005403.2
XM_011538675.2 1046 Intron XP_011536977.1
XM_011538676.2 1046 Intron XP_011536978.1
XM_011538677.2 1046 Intron XP_011536979.1
XM_011538678.2 1046 Intron XP_011536980.1
Gene
STAC3
Gene Name
SH3 and cysteine rich domain 3
There are no transcripts associated with this gene.

View Full Product Details