Product Details

SNP ID
rs202098511
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54645639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGATCTCTGCTTCCTTGGCTTTG[A/G]TGCCTGTCTGGCTAACCCTGGGTCT
Phenotype
MIM: 606634
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCD PubMed Links

Gene Details

Gene
DCD
Gene Name
dermcidin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300854.1 356 Missense Mutation CCA,TCA P56S NP_001287783.1
NM_053283.3 356 Missense Mutation CCA,TCA P56S NP_444513.1

View Full Product Details