Product Details

SNP ID
rs201243887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85017511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAACCATTTTCTTAAAGTTGAC[C/T]TTGTGCAAGAACATGGCACCTGTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSPAN19 PubMed Links

Gene Details

Gene
TSPAN19
Gene Name
tetraspanin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100917.1 578 Missense Mutation AAG,AGG K180R NP_001094387.1
XM_005268669.4 578 Missense Mutation AAG,AGG K180R XP_005268726.1
XM_017018862.1 578 Missense Mutation AAG,AGG K180R XP_016874351.1
XM_017018863.1 578 Missense Mutation AAG,AGG K178R XP_016874352.1
XM_017018864.1 578 Missense Mutation AAG,AGG K180R XP_016874353.1

View Full Product Details