Product Details

SNP ID
rs201446181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57758170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTGCTGCCATAGTTCTGGGCTC[A/G]CTCTTCCTGGTTGCCAGCATCTCCT
Phenotype
MIM: 123829 MIM: 609506 MIM: 613336 MIM: 181035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDK4 PubMed Links

Gene Details

Gene
CDK4
Gene Name
cyclin dependent kinase 4
There are no transcripts associated with this gene.

Gene
CYP27B1
Gene Name
cytochrome P450 family 27 subfamily B member 1
There are no transcripts associated with this gene.

Gene
MARCH9
Gene Name
membrane associated ring-CH-type finger 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138396.5 1007 Silent Mutation TCA,TCG S192S NP_612405.2
Gene
MIR6759
Gene Name
microRNA 6759
There are no transcripts associated with this gene.

Gene
TSPAN31
Gene Name
tetraspanin 31
There are no transcripts associated with this gene.

View Full Product Details