Product Details

SNP ID
rs201304555
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56952048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGCTTCATAGAGCCTTGGCCGGG[C/T]TTGGAGAGACCCAGTACATAATGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RDH16 PubMed Links

Gene Details

Gene
RDH16
Gene Name
retinol dehydrogenase 16 (all-trans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320108.1 2847 Missense Mutation AAC,AGC N167S NP_001307037.1
NM_003708.4 2847 Missense Mutation AAC,AGC N312S NP_003699.3

View Full Product Details